missing translation for 'onlineSavingsMsg'
Learn More

Amoxicillin Related Compound F, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023010
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023010 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023010 Supplier Merck Emd Millipore Supplier No. PHR2193100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 126247-63-0
Flash Point Not Applicable
Molecular Formula C10H8N2O2
Linear Formula C10H8N2O2
Quantity 100 mg
Synonym 3-(4-Hydroxyphenyl)pyrazin-2-ol
Molecular Weight (g/mol) 188.18
Formula Weight 188.18
Grade Certified Reference Material
Physical Form Powder