missing translation for 'onlineSavingsMsg'
Learn More

Amoxicillin Crystalline MP Biomedicals

Catalog No. p-3178634
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
0219014505 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0219014505 Supplier MP Biomedicals Inc Supplier No. 0219014505

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26787-78-0
Molecular Formula C16H19N3O5S
Molecular Weight (g/mol) 365.404
InChI Key LSQZJLSUYDQPKJ-NJBDSQKTSA-N
Synonym Amoxycillin, Amolin, AMPC
PubChem CID 33613
ChEBI CHEBI:2676
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)C(=O)O)C

Specifications

Type Amoxicillin
Physical Form Crystalline
Melting Point 194°C
Sterility Non-sterile
Color White
Quantity 5 g
Solubility Information 10.7 μg/mL
IUPAC Name (2 S,5 R,6 R)-6-[[(2 R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0 ]heptane-2-carboxylic acid
Formula Weight 365.4
Grade Not Available
Chemical Name or Material Amoxicillin
Show More Show Less