missing translation for 'onlineSavingsMsg'
Learn More

Amoxapine 97.0+%, TCI America™

Catalog No. p-7117944
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A24991G Supplier TCI America Supplier No. A24991G
Only null left

Chemical Identifiers

CAS 14028-44-5
Molecular Formula C17H16ClN3O
Molecular Weight (g/mol) 313.785
MDL Number MFCD00069210
InChI Key QWGDMFLQWFTERH-UHFFFAOYSA-N
Synonym amoxapine, asendin, demolox, moxadil, amoxan, amoxapinum, amoxapina, amoxepine, asendis, desmethylloxapin
PubChem CID 2170
ChEBI CHEBI:2675
IUPAC Name 8-chloro-6-piperazin-1-ylbenzo[b][1,4]benzoxazepine
SMILES C1CN(CCN1)C2=NC3=CC=CC=C3OC4=C2C=C(C=C4)Cl

Specifications

Melting Point 179°C
Color White-Yellow
Quantity 1 g
Formula Weight 313.79
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Amoxapine
RTECSNumber HQ4025500
TSCA No