Learn More
Ammonium vanadium oxide, Puratronic™, 99.995% (metals basis)
Description
It is used as a catalyst in organic synthesis, and as an analytical reagent in laboratories. It is employed in the preparation of vanadate. It acts as a sensor for the determination of N-acetyltransferase activity in humans. Further it finds use in dyes, varnishes, indelible inks, drier for paints, and in photography. With copper carbonate, it has been shown to enhance the catalytic activities. Ammonium metavanadate is an environmental-friendly catalyst to prepare alpha-hydroxyphosphonate derivatives in high yield through the reaction of aryl/heteroaryl aldehydes with triethylphosphonate derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 200°C (decomposition) |
| Color | White |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 99.995% (Metals basis) |
| Solubility Information | Slightly soluble in water. Soluble in dilute ammonia and diethanolamine. Insoluble in ether,and alcohol. |
| Formula Weight | 116.98 |
| Density | 2.33 |
| Chemical Name or Material | Ammonium vanadium oxide |
Safety and Handling
| Hazard Category | H301-H319-H332-H361d-H372 |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P304+P340-P305+P351+P338-P308+P313-P330-P501c |
| DOTInformation | DOT Class: 6.1, Packaging Group: II |
| EINECSNumber | 232-261-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only