missing translation for 'onlineSavingsMsg'
Learn More

Ammonium vanadium oxide, Puratronic™, 99.995% (metals basis)

Catalog No. AA1090214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1090214 25 g
AA1090206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1090214 Supplier Thermo Scientific Chemicals Supplier No. 01090214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7803-55-6 | H4NO3V | 116.98 g/mol

It is used as a catalyst in organic synthesis, and as an analytical reagent in laboratories. It is employed in the preparation of vanadate. It acts as a sensor for the determination of N-acetyltransferase activity in humans. Further it finds use in dyes, varnishes, indelible inks, drier for paints, and in photography. With copper carbonate, it has been shown to enhance the catalytic activities. Ammonium metavanadate is an environmental-friendly catalyst to prepare alpha-hydroxyphosphonate derivatives in high yield through the reaction of aryl/heteroaryl aldehydes with triethylphosphonate derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7803-55-6
Molecular Formula H4NO3V
Molecular Weight (g/mol) 116.98
MDL Number MFCD00011430
InChI Key YBVKNHXQSRDWAA-UHFFFAOYSA-M
Synonym ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
PubChem CID 516859
IUPAC Name vanadiumoylol amine
SMILES N.O[V](=O)=O

Specifications

Melting Point 200°C (decomposition)
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.995% (Metals basis)
Solubility Information Slightly soluble in water. Soluble in dilute ammonia and diethanolamine. Insoluble in ether,and alcohol.
Formula Weight 116.98
Density 2.33
Chemical Name or Material Ammonium vanadium oxide
Hazard Category H301-H319-H332-H361d-H372
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P304+P340-P305+P351+P338-P308+P313-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: II
EINECSNumber 232-261-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only