missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tungsten oxide hydrate

CAS: 12333-11-8 | (NH4)6W12O39·xH2O

$92.15 - $591.38

Chemical Identifiers

CAS 12333-11-8
Molecular Formula (NH4)6W12O39·xH2O
MDL Number MFCD00042673
Synonym Ammonium metatungstate hydrate
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4479214
View Documents
Thermo Scientific Chemicals
04479214
25 g
Each for $92.15
Only null left
 
AA4479222
View Documents
Thermo Scientific Chemicals
04479222
100 g
Each for $178.30
Only null left
 
AA4479236
View Documents
Thermo Scientific Chemicals
04479236
500 g
Each for $591.38
Only null left
 
Description

Description

Ammonium tungsten oxide hydrate is widely utilized as a raw material for the synthesis of tungsten catalysts, which is used in a variety of reactions like oxidation, hydroxylation, hydrogenation and polymerization. It is used as a reagent for chemical analysis like medical diagnosis. It is used as a precursor material for the preparation of high-purity WO3 which is used as thin-film substrate for certain semiconductor devices. It acts as a precursor to a variety of POM (polyoxometalates).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12333-11-8
(NH4)6W12O39·xH2O
MFCD00042673
Ammonium metatungstate hydrate

Specifications

12333-11-8
25 g
MFCD00042673
Soluble in water. Insoluble in alcohol.
Ammonium tungsten oxide hydrate
Powder
(NH4)6W12O39·xH2O
Ammonium metatungstate hydrate
2938.41 (Anhydrous)
Videos
Safety and Handling

Safety and Handling

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 683-059-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only