missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Trifluoroacetate, Honeywell Fluka™

for HPLC,≥99.0% (NT)

$2163.66 - $2163.66

Chemical Identifiers

CAS 3336-58-1
Molecular Formula C2H5F3NO2
Molecular Weight (g/mol) 132.06
MDL Number MFCD00012615,MFCD03095537
InChI Key YCNIBOIOWCTRCL-UHFFFAOYSA-O
Synonym ammonium trifluoroacetate, azanium 2,2,2-trifluoroacetate, ammoniumtrifluoroacetate, ammonium triflouroacetate, cf3coonh4, acmc-1cq68, 2,2,2-trifluoroacetic acid, azamethane, acetic acid, trifluoro-, ammonium salt 8ci 9ci
PubChem CID 86601334
IUPAC Name azanium;2,2,2-trifluoroacetic acid
SMILES [NH4+].OC(=O)C(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6002030
View Documents
Honeywell Chemicals
1783950G
50 g
Each for $2,163.66
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

3336-58-1
132.06
YCNIBOIOWCTRCL-UHFFFAOYSA-O
86601334
[NH4+].OC(=O)C(F)(F)F
C2H5F3NO2
MFCD00012615,MFCD03095537
ammonium trifluoroacetate, azanium 2,2,2-trifluoroacetate, ammoniumtrifluoroacetate, ammonium triflouroacetate, cf3coonh4, acmc-1cq68, 2,2,2-trifluoroacetic acid, azamethane, acetic acid, trifluoro-, ammonium salt 8ci 9ci
azanium;2,2,2-trifluoroacetic acid

Specifications

3336-58-1
C2H5F3NO2
MFCD00012615,MFCD03095537
ammonium trifluoroacetate, azanium 2,2,2-trifluoroacetate, ammoniumtrifluoroacetate, ammonium triflouroacetate, cf3coonh4, acmc-1cq68, 2,2,2-trifluoroacetic acid, azamethane, acetic acid, trifluoro-, ammonium salt 8ci 9ci
YCNIBOIOWCTRCL-UHFFFAOYSA-O
azanium;2,2,2-trifluoroacetic acid
86601334
≥99% (NT)
50 g
CF3CO2NH4
NONH for all modes of transport
Soluble in water 0.1g/mL at 25°C
[NH4+].OC(=O)C(F)(F)F
132.06
131.05g/mol
Ammonium Trifluoroacetate
Videos
Safety and Handling

Safety and Handling

P280-P304 + P340 + P312-P305 + P351 + P338-P337 + P313

H315-H319-H335

EINECSNumber : 222-072-4

SDS
Documents

Documents