missing translation for 'onlineSavingsMsg'
Learn More

Ammonium trifluoroacetate, 98%

Catalog No. p-7029141
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2657714 25 g
AAH2657722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2657714 Supplier Thermo Scientific Chemicals Supplier No. H2657714
Only null left

CAS: 3336-58-1 | C2H5F3NO2 | 132.06 g/mol

Used in the synthesis of hexafluoro-2-aminopentan-4-one ligand and in tuning and calibrating new liquid chromatography/mass spectrometry systems. Applications of ammonium trifluoroacetate as an additive for elution of chiral acids and bases from derivatized polysaccharide stationary phases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of hexafluoro-2-aminopentan-4-one ligand and in tuning and calibrating new liquid chromatography/mass spectrometry systems. Applications of ammonium trifluoroacetate as an additive for elution of chiral acids and bases from derivatized polysaccharide stationary phases

Solubility
Soluble in Water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture. Keep under dry inert gas.

Chemical Identifiers

CAS 3336-58-1
Molecular Formula C2H5F3NO2
Molecular Weight (g/mol) 132.06
MDL Number MFCD00012615,MFCD03095537
InChI Key YCNIBOIOWCTRCL-UHFFFAOYSA-O
Synonym ammonium trifluoroacetate, azanium 2,2,2-trifluoroacetate, ammoniumtrifluoroacetate, ammonium triflouroacetate, cf3coonh4, acmc-1cq68, 2,2,2-trifluoroacetic acid, azamethane, acetic acid, trifluoro-, ammonium salt 8ci 9ci
PubChem CID 86601334
IUPAC Name azanium;2,2,2-trifluoroacetic acid
SMILES [NH4+].OC(=O)C(F)(F)F

Specifications

Melting Point 123°C to 126°C
Linear Formula CF3COONH4
Quantity 25 g
Sensitivity Hygroscopic
Solubility Information Soluble in Water.
Formula Weight 131.05
Percent Purity 98%
Chemical Name or Material Ammonium trifluoroacetate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-072-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only