missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. p-31037
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A709500 500 g Poly Bottle
A7093 3 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A709500 Supplier Thermo Fisher Scientific Supplier No. FLA709500
Only null left

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
MDL Number MFCD00011428
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Melting Point 149°C
Color White
pH 5 to 6
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥97.5 %
Linear Formula NH4SCN
Formula Weight 76.12
Percent Purity ≥97.5%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Density 1.300g/cm³
Chemical Name or Material Ammonium Thiocyanate
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Contact with acids liberates toxic gas. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use only under a chemical fume hood. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:2
Flammability:1
Instability:1