missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Thiocyanate, Certified, 0.100N ±0.001N (0.1M), LabChem™
$87.11 - $256.93
Chemical Identifiers
| CAS | 1762-95-4 |
|---|---|
| Molecular Formula | CH4N2S |
| Molecular Weight (g/mol) | 76.117 |
| InChI Key | SOIFLUNRINLCBN-UHFFFAOYSA-N |
| Synonym | ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt |
| PubChem CID | 15666 |
| IUPAC Name | azanium;thiocyanate |
| SMILES | C(#N)[S-].[NH4+] |
Chemical Identifiers
| 1762-95-4 | |
| 76.117 | |
| ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt | |
| azanium;thiocyanate |
| CH4N2S | |
| SOIFLUNRINLCBN-UHFFFAOYSA-N | |
| 15666 | |
| C(#N)[S-].[NH4+] |
Specifications
| 1762-95-4,7732-18-5 | |
| 99.24,0.76 | |
| 1 L | |
| NH4SCN | |
| Soluble in water | |
| C(#N)[S-].[NH4+] | |
| 76.117 | |
| 76.12 | |
| 0.100N ±0.001N (0.1M) | |
| Poly Bottle | |
| Ammonium Thiocyanate |
| Colorless | |
| Liquid | |
| CH4N2S | |
| ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt | |
| SOIFLUNRINLCBN-UHFFFAOYSA-N | |
| azanium;thiocyanate | |
| 15666 | |
| Certified | |
| Passes Test | |
| Gaseous Ammonia; Sulfur compounds; Nitrogen oxides; Carbon monoxide; Carbon dioxide |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature