missing translation for 'onlineSavingsMsg'
Learn More

Ammonium thiocyanate, 98%, ACS reagent

Catalog No. p-45678
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423415000 500 g Plastic bottle
AC423410030 3 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC423415000 Supplier Thermo Scientific Chemicals Supplier No. 42341 5000
Only null left

CAS: 1762-95-4 | CH4N2S | 76.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.12
MDL Number MFCD00011428
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Melting Point 149.0°C
Color Colorless or White
pH 4.5 to 6.0 (5% soln. at 25°C)
Physical Form Crystals
Quantity 500 g
Assay Percent Range 98%
Linear Formula NH4SCN
Merck Index 15, 558
Solubility Information Solubility in water: 1600g/L (20°C). Other solubilities: soluble in alcohol, acetone and ammonia, practically insoluble in chcl3 and ethyl acetate
Formula Weight 76.12
Percent Purity 98%
Grade ACS Reagent
Packaging Plastic bottle
Residue after Ignition 0.025% max
Specific Gravity 1.3
Density 1.3000g/mL
Chemical Name or Material Ammonium thiocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
Contact with acids liberates very toxic gas.
Precautionary Statement GHS P Statement
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF
EINECSNumber 217-175-6
RTECSNumber XK7875000
TSCA TSCA

RUO – Research Use Only