missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium tetrathiomolybdate, 99.95%, (trace metal basis)
CAS: 15060-55-6 | H10MoN2S4 | 262.28 g/mol
$85.25 - $1,332.03
Chemical Identifiers
| CAS | 15060-55-6 |
|---|---|
| Molecular Formula | H10MoN2S4 |
| Molecular Weight (g/mol) | 262.28 |
| MDL Number | MFCD00136013 |
| InChI Key | ZKKLPDLKUGTPME-UHFFFAOYSA-N |
| Synonym | ammonium tetrathiomolybdate, molybdate 2-, tetrathioxo-, ammonium 1:2 , t-4, tiomolibdate diammonium, unii-4v6i63lw1e, ammonium thiomolybdate, molybdate 2-, tetrathioxo-, diammonium, t-4, ammonium tetrathiomolybdate vi, molybdate 2-, tetrathioxo-, diammonium, beta-4 |
| PubChem CID | 15251598 |
| SMILES | [NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC389530010
|
Thermo Scientific Chemicals
389530010 |
1 g | Glass bottle |
N/A
|
|
||||
|
AC389530100
|
Thermo Scientific Chemicals
389530100 |
10 g | Glass bottle |
N/A
|
|
||||
|
AC389530500
|
Thermo Scientific Chemicals
389530500 |
50 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
| 15060-55-6 | |
| 262.28 | |
| ZKKLPDLKUGTPME-UHFFFAOYSA-N | |
| 15251598 |
| H10MoN2S4 | |
| MFCD00136013 | |
| ammonium tetrathiomolybdate, molybdate 2-, tetrathioxo-, ammonium 1:2 , t-4, tiomolibdate diammonium, unii-4v6i63lw1e, ammonium thiomolybdate, molybdate 2-, tetrathioxo-, diammonium, t-4, ammonium tetrathiomolybdate vi, molybdate 2-, tetrathioxo-, diammonium, beta-4 | |
| [NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S |
Specifications
| 300.0°C | |
| 15060-55-6 | |
| 1 g | |
| H10MoN2S4 | |
| MFCD00136013 | |
| ammonium tetrathiomolybdate, molybdate 2-, tetrathioxo-, ammonium 1:2 , t-4, tiomolibdate diammonium, unii-4v6i63lw1e, ammonium thiomolybdate, molybdate 2-, tetrathioxo-, diammonium, t-4, ammonium tetrathiomolybdate vi, molybdate 2-, tetrathioxo-, diammonium, beta-4 | |
| [NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S | |
| 262.28 | |
| 260.28 | |
| Trace Metal Basis | |
| Ammonium tetrathiomolybdate |
| White | |
| Crystals or Powder | |
| 99.95% | |
| (NH4)2MoS4 | |
| 15,557 | |
| ZKKLPDLKUGTPME-UHFFFAOYSA-N | |
| diazanium;bis(sulfanylidene)molybdenum;sulfanide | |
| 15251598 | |
| 99.95% | |
| Glass bottle |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only