Learn More
Ammonium tetrathiomolybdate, 99.95% (metals basis)
CAS: 15060-55-6 | H10MoN2S4 | 262.28 g/mol
$236.55 - $930.32
Chemical Identifiers
| CAS | 15060-55-6 |
|---|---|
| Molecular Formula | H10MoN2S4 |
| Molecular Weight (g/mol) | 262.28 |
| MDL Number | MFCD00136013 |
| InChI Key | ZKKLPDLKUGTPME-UHFFFAOYSA-N |
| Synonym | ammonium tetrathiomolybdate, molybdate 2-, tetrathioxo-, ammonium 1:2 , t-4, tiomolibdate diammonium, unii-4v6i63lw1e, ammonium thiomolybdate, molybdate 2-, tetrathioxo-, diammonium, t-4, ammonium tetrathiomolybdate vi, molybdate 2-, tetrathioxo-, diammonium, beta-4 |
| PubChem CID | 15251598 |
| IUPAC Name | diazanium;bis(sulfanylidene)molybdenum;sulfanide |
| SMILES | [NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S |
Description
Ammonium tetrathiomolybdate is used as an important reagent in synthetic chemistry and as a building block in bioinorganic chemistry. It is used in the treatment of copper toxicosis in animals and in Wilson's disease. It acts as a chelator in vivo to reduce copper levels. It interferes with angiogenesis and reduces tumor growth. Further, it is used in the synthesis of heterometallic sulfide clusters stabilized with dppm [bis(diphenylphosphino)methane] ligands. It is involved in the ring opening of aziridines leading to sulfides and disulfides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 15060-55-6 | |
| 262.28 | |
| ZKKLPDLKUGTPME-UHFFFAOYSA-N | |
| 15251598 | |
| [NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S |
| H10MoN2S4 | |
| MFCD00136013 | |
| ammonium tetrathiomolybdate, molybdate 2-, tetrathioxo-, ammonium 1:2 , t-4, tiomolibdate diammonium, unii-4v6i63lw1e, ammonium thiomolybdate, molybdate 2-, tetrathioxo-, diammonium, t-4, ammonium tetrathiomolybdate vi, molybdate 2-, tetrathioxo-, diammonium, beta-4 | |
| diazanium;bis(sulfanylidene)molybdenum;sulfanide |
Specifications
| 15060-55-6 | |
| 5 g | |
| H10MoN2S4 | |
| ammonium tetrathiomolybdate, molybdate 2-, tetrathioxo-, ammonium 1:2 , t-4, tiomolibdate diammonium, unii-4v6i63lw1e, ammonium thiomolybdate, molybdate 2-, tetrathioxo-, diammonium, t-4, ammonium tetrathiomolybdate vi, molybdate 2-, tetrathioxo-, diammonium, beta-4 | |
| ZKKLPDLKUGTPME-UHFFFAOYSA-N | |
| diazanium;bis(sulfanylidene)molybdenum;sulfanide | |
| 15251598 | |
| Hygroscopic |
| Crystalline | |
| 99.95% (metals basis) | |
| MFCD00136013 | |
| Slightly soluble in water. | |
| [NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S | |
| 262.28 | |
| 260.27 | |
| Ammonium tetrathiomolybdate |
Safety and Handling
P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : QA4668250
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only