missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tetrafluoroborate, 99%, pure

Catalog No. AC310965000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC310965000 500 g Plastic bottle
AC310961000 100 g Plastic bottle
AC310960025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC310965000 Supplier Thermo Scientific Chemicals Supplier No. 310965000

CAS: 13826-83-0 | BF4H4N | 104.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13826-83-0
Molecular Formula BF4H4N
Molecular Weight (g/mol) 104.84
MDL Number MFCD06245785
InChI Key PDTKOBRZPAIMRD-UHFFFAOYSA-O
Synonym ammonium tetrafluoroborate, ammonium fluoroborate, ammonium fluoborate, ammonium borofluoride, unii-c945z80o8x, ammonium tetrafluroborate, azanium tetrafluoroborate, arnmonium tetrafluoroborate, nh4bf4, ksc492c2p
PubChem CID 9964072
SMILES [NH4+].F[B-](F)(F)F

Specifications

Melting Point 230.0°C
Color Colorless
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99%
Linear Formula NH4BF4
Fieser 05,17; 06,132
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ammonium hydride
IUPAC Name azanium;tetrafluoroborate
Formula Weight 104.84
Percent Purity 99%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Ammonium tetrafluoroborate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 237-531-4

RUO – Research Use Only