missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tetrafluoroborate, 97%

Catalog No. AA1148836
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1148836 500 g
AA11488A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1148836 Supplier Thermo Scientific Chemicals Supplier No. 01148836

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13826-83-0 | BF4H4N | 104.84 g/mol

Ammonium fluoroborate is used as an analytical reagent. In the textile printing and dyeing industry, it used as a catalyst for resin finishing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13826-83-0
Molecular Formula BF4H4N
Molecular Weight (g/mol) 104.84
MDL Number MFCD06245785
InChI Key PDTKOBRZPAIMRD-UHFFFAOYSA-O
Synonym ammonium tetrafluoroborate, ammonium fluoroborate, ammonium fluoborate, ammonium borofluoride, unii-c945z80o8x, ammonium tetrafluroborate, azanium tetrafluoroborate, arnmonium tetrafluoroborate, nh4bf4, ksc492c2p
PubChem CID 9964072
SMILES [NH4+].F[B-](F)(F)F

Specifications

Melting Point 220°C (sublimation)
Color White
Physical Form Powder
Quantity 500 g
UN Number UN3260
Solubility Information Soluble in water.
IUPAC Name azanium;tetrafluoroborate
Formula Weight 104.84
Percent Purity 97%
Odor Ammonia-like
Sensitivity Hygroscopic
Density 1.871 g/mL
Chemical Name or Material Ammonium tetrafluoroborate
Show More Show Less
Hazard Category H314-H318-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 237-531-4
RTECSNumber BQ6100000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only