Learn More
Ammonium tetrachloropalladate(II), Pd 36% min
CAS: 13820-40-1 | Cl4H8N2Pd | 284.30 g/mol
$227.73 - $227.73
Chemical Identifiers
| CAS | 13820-40-1 |
|---|---|
| Molecular Formula | Cl4H8N2Pd |
| Molecular Weight (g/mol) | 284.30 |
| MDL Number | MFCD00010884 |
| InChI Key | IRKVTUVFHGUMMN-UHFFFAOYSA-L |
| Synonym | ammonium tetrachloropalladate ii, ammonium palladium chloride, diammonium tetrachloropalladate 2-, ammonium chloropalladite, bisammonium tetrachloropalladate 2-, diammonium tetrachloro palladite, ammonium tetrachloropalladate, ccris 7828, palladate 2-, tetrachloro-, diammonium, sp-4-1, ammoniumpalladiumchloride |
| PubChem CID | 61682 |
| ChEBI | CHEBI:60149 |
| IUPAC Name | diazanium;tetrachloropalladium(2-) |
| SMILES | [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pd++] |
Description
It is used as catalyst, and as a standard for spectral analysis. It reacts with creatinine to form various Pd-creatinine complexes which are used in semiconductors, catalysis, mixed oxidation state studies, and electron conductivity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13820-40-1 | |
| 284.30 | |
| IRKVTUVFHGUMMN-UHFFFAOYSA-L | |
| 61682 | |
| diazanium;tetrachloropalladium(2-) |
| Cl4H8N2Pd | |
| MFCD00010884 | |
| ammonium tetrachloropalladate ii, ammonium palladium chloride, diammonium tetrachloropalladate 2-, ammonium chloropalladite, bisammonium tetrachloropalladate 2-, diammonium tetrachloro palladite, ammonium tetrachloropalladate, ccris 7828, palladate 2-, tetrachloro-, diammonium, sp-4-1, ammoniumpalladiumchloride | |
| CHEBI:60149 | |
| [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pd++] |
Specifications
| (decomposition) | |
| Brown to Green | |
| 1 g | |
| (NH4)2PdCl4 | |
| ammonium tetrachloropalladate ii, ammonium palladium chloride, diammonium tetrachloropalladate 2-, ammonium chloropalladite, bisammonium tetrachloropalladate 2-, diammonium tetrachloro palladite, ammonium tetrachloropalladate, ccris 7828, palladate 2-, tetrachloro-, diammonium, sp-4-1, ammoniumpalladiumchloride | |
| IRKVTUVFHGUMMN-UHFFFAOYSA-L | |
| diazanium;tetrachloropalladium(2-) | |
| 61682 | |
| 284.31 | |
| Hygroscopic | |
| Ammonium tetrachloropalladate(II) |
| 13820-40-1 | |
| Powder | |
| Cl4H8N2Pd | |
| MFCD00010884 | |
| Soluble in water. Insoluble in ethanol. | |
| [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pd++] | |
| 284.30 | |
| CHEBI:60149 | |
| Pd 36% min. | |
| 2.17 g/mL |
Safety and Handling
GHS H Statement
H318-H302-H315
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
EINECSNumber : 237-498-6
RTECSNumber : BP5400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only