missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tartrate, 98.8%, MP Biomedicals™

$347.77 - $347.77

Chemical Identifiers

CAS 3164-29-2
Molecular Formula C4H12N2O6
Molecular Weight (g/mol) 184.15
MDL Number MFCD00013073
InChI Key NGPGDYLVALNKEG-OLXYHTOASA-N
Synonym ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
PubChem CID 13652278
ChEBI CHEBI:63075
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid diamine
SMILES N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19504290
SDS View Documents
MP Biomedicals Inc
0219504290
500 g
Each for $347.77
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3164-29-2
184.15
NGPGDYLVALNKEG-OLXYHTOASA-N
13652278
(2R,3R)-2,3-dihydroxybutanedioic acid diamine
C4H12N2O6
MFCD00013073
ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
CHEBI:63075
N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O

Specifications

White
Crystalline Powder
0.988
MFCD00013073
NGPGDYLVALNKEG-OLXYHTOASA-N
(2R,3R)-2,3-dihydroxybutanedioic acid diamine
13652278
184.1
Ammonium tartrate
3164-29-2
500 g
C4H12N2O6
ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O
184.15
CHEBI:63075
1.601g/mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'