missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, Ultrapure Bioreagent, J.T. Baker™

Catalog No. p-5390693
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002323 1 kg Poly Bottle
02002324 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02002323 Supplier Avantor Supplier No. 402702
Only null left

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Protein Precipitation, Liquid Chromatography and Molecular Biology Applications

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name diazanium;sulfate
SMILES N.N.OS(O)(=O)=O

Specifications

Quantity 1 kg
Formula Weight 132.14
Grade Ultrapure Bioreagent
Packaging Poly Bottle
Specific Gravity 1L = 1.77kg
Chemical Name or Material Ammonium Sulfate