missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, Ultra Pure, 1kg, MP Biomedicals

Catalog No. ICN808211
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN808211 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN808211 Supplier MP Biomedicals Inc Supplier No. 04808211
Only null left

  • Fe: <<0.5 ppm
  • Mg: <<0.13 ppm
  • Pb: <<0.1 ppm
  • Mn: <<0.06 ppm
  • Ca: <<0.1 ppm
  • Zn: <<0.5 ppm
  • Cu: <<0.21 ppm
  • O.D. (260nm) of a saturated solution: <<0.1

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O

Specifications

Quantity 1 kg
Grade Ultrapure
Chemical Name or Material Ammonium Sulfate, Ultra Pure