missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium sulfate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99%
$391.58 - $4133.63
Chemical Identifiers
| CAS | 7783-20-2 |
|---|---|
| Molecular Formula | H8N2O4S |
| Molecular Weight (g/mol) | 132.13 |
| MDL Number | MFCD00003391 |
| InChI Key | BFNBIHQBYMNNAN-UHFFFAOYSA-N |
| Synonym | ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate |
| PubChem CID | 6097028 |
| ChEBI | CHEBI:62946 |
| IUPAC Name | diazanium;sulfate |
| SMILES | N.N.OS(O)(=O)=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7783-20-2 | |
| 132.13 | |
| BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
| 6097028 | |
| diazanium;sulfate |
| H8N2O4S | |
| MFCD00003391 | |
| ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
| CHEBI:62946 | |
| N.N.OS(O)(=O)=O |
Specifications
| 7783-20-2 | |
| H8N2O4S | |
| MFCD00003391 | |
| ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
| N.N.OS(O)(=O)=O | |
| 132.13 | |
| CHEBI:62946 | |
| ≥99% | |
| ≤0.005% (as SO4) | |
| Ammonium Sulfate, Reag. Ph. Eur. reag. ISO |
| 1 kg | |
| (NH4)2SO4 | |
| NONH for all modes of transport | |
| BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
| diazanium;sulfate | |
| 6097028 | |
| 132.14g/mol | |
| ACS/puriss. p.a. | |
| 1.77g/mL at 25°C |
Safety and Handling
EINECSNumber : 231-984-1
RTECSNumber : BS4500000