missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, Granular, Purified, Spectrum™ Chemical

Catalog No. 18608786
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608786 25 kg Poly Pail
18608784 500 g Poly Pail
18608782 2.5 kg Poly Pail
18608783 12 kg Poly Pail
18608785 50 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608786 Supplier Spectrum Chemical Mfg Cor Supplier No. A124725KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7783-20-2

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 25 kg
Molecular Weight (g/mol) 132.14
Grade Purified
Packaging Poly Pail
Chemical Name or Material Ammonium Sulfate, Granular

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.