missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate (Granular/Certified ACS), Fisher Chemical™

Catalog No. p-31017
Change view
Click to view available options
Quantity:
500 g
3 kg
10 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A702500 500 g Poly Bottle
A7023 3 kg Poly Bottle
A70210 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A702500 Supplier Thermo Fisher Scientific Supplier No. FLA702500
Only null left

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O

Specifications

Melting Point 280°C
Color Colorless
pH 5 to 6
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥99.0 %
Linear Formula (NH4)2SO4
Formula Weight 132.14
Percent Purity ≥99.0%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Density 1.77g/mL;ph
Chemical Name or Material Ammonium Sulfate
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:1
Flammability:0
Instability:0