missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sulfate, Fisher BioReagents
Description
This enzyme-grade Ammonium Sulfate is suitable for use in the purification of proteins.
Specifications
Specifications
| Melting Point | 280°C |
| Color | Colorless |
| pH | 5 to 6 |
| Physical Form | Solid |
| Quantity | 1 kg |
| Linear Formula | (NH4)2SO4 |
| Merck Index | 15, 552 |
| Formula Weight | 132.14 |
| Percent Purity | ≥99.5% |
| Identification | Pass Test |
| Show More |
Safety and Handling
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause skin, eye, and respiratory tract irritation. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . . NFPA Health:1 Flammability:0 Instability:0 |
| Recommended Storage | RT |