missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical

A1245, 7783-20-2, (NH4)2SO4

$266.21 - $6595.46

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O
View More Specs

Products 9
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18600656
View Documents
Spectrum Chemical Mfg Cor
A1245125GM
125 g Poly Bottle
Each for $266.21
Only null left
 
18600657
View Documents
Spectrum Chemical Mfg Cor
A1245500GM
500 g Poly Bottle
Each for $403.14
Only null left
 
18600659
missing translation for 'viewDocuments'
Spectrum Chemical Mfg Cor
A124525KG
2.5 kg Poly Bottle
Each for $1,003.36
Only null left
 
18600658
missing translation for 'viewDocuments'
Spectrum Chemical Mfg Cor
A1245500GMCS6
6 x 500 g Poly Bottle
Case of 6 Each for $1,424.36
Only null left
 
18600660
missing translation for 'viewDocuments'
Spectrum Chemical Mfg Cor
A124525KGCS4
4 x 2.5 kg Poly Bottle
Case of 4 Each for $3,294.93
Only null left
 
18600661
missing translation for 'viewDocuments'
Spectrum Chemical Mfg Cor
A124512KG
12 kg Poly Pail
Each for $2,178.96
Only null left
 
18600664
missing translation for 'viewDocuments'
Spectrum Chemical Mfg Cor
A124525KGBL
25 kg Fiber Drum
Each for $3,845.50
Only null left
 
18600662
missing translation for 'viewDocuments'
Spectrum Chemical Mfg Cor
A124550KGBL
50 kg Fiber Drum
Each for $4,547.46
Only null left
 
18600663
missing translation for 'viewDocuments'
Spectrum Chemical Mfg Cor
A124550KGBLPL
50 kg Poly Drum
Each for $6,595.46
Only null left
 
Description

Description

Spectrum™ Chemical Ammonium Sulfate, Crystal, Reagent, ACS is an inorganic salt that is very soluble in water Most often used as a soil fertilizer to release minute amounts of acid. It is also used in biochemistry as a precipitant to purify protein, used as a food additive and flame retardant. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.
Specifications

Chemical Identifiers

7783-20-2
132.13
BFNBIHQBYMNNAN-UHFFFAOYSA-N
N.N.OS(O)(=O)=O
H8N2O4S
MFCD00003391
sulfuric acid diamine

Specifications

7783-20-2
5.0-6.0
H8N2O4S
BFNBIHQBYMNNAN-UHFFFAOYSA-N
sulfuric acid diamine
99%
Poly Bottle
100%
125 g
MFCD00003391
N.N.OS(O)(=O)=O
132.13
ACS
0.005%
Videos
SDS
Documents

Documents