missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, ACS Grade, ≥ 99.0%, LabChem™

Catalog No. LC112981
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC112981 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC112981 Supplier LabChem Supplier No. LC112981

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O

Specifications

Melting Point 280°C
Color White
CAS Max % 100
Physical Form Granules
Quantity 500 g
Linear Formula (NH4)2SO4
Solubility Information Soluble in water
Formula Weight 132.13
Percent Purity ≥99.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.77g/mL
Decomposition Information Gaseous Ammonia; Sulfur compounds
Density 1.77g/mL
Chemical Name or Material Ammonium Sulfate
Show More Show Less
Hazard Statement GHS H Statement
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 231-984-1
Recommended Storage Room Temperature