missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfate, ACS, 99.0% min

Catalog No. p-3256890
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
10 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1156622 100 g
AA1156636 500 g
AA11566A3 2 kg
AA11566A9 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1156622 Supplier Thermo Scientific Chemicals Supplier No. 01156622
Only null left

In galvanizing steel, manufacturing of ammonia alum and sulfuric acid | CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol

In biochemistry it is used for purification of proteins via ammonium sulfate precipitation method. Ammonium sulfate is used in agriculture as fertilizer. It is a slow releasing nitrogenous fertilizer and a rich source of sulphur nutrient. It is employed as a dough conditioner in bread products. It finds use as raw materials for various chemical products such as dyes and pharmacetical products. It is a component of flame-proofing agents and fire extinguisher powder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
SMILES N.N.OS(O)(=O)=O

Specifications

Melting Point ∽280°C (decomposition)
Color White
Physical Form Granules
Quantity 100 g
Assay Percent Range 99.0%
Solubility Information Highly soluble in water. It is insoluble in acetone,ether and ethanol. It is also insoluble in liquid ammonia.
IUPAC Name sulfuric acid diamine
Formula Weight 132.14
Grade ACS
Density 1.769 g/mL
Chemical Name or Material Ammonium sulfate, For analysis ACS
Show More Show Less
Hazard Category H303
EINECSNumber 231-984-1
RTECSNumber BS4500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only