missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate,≥99%, ReagentPlus™,, Honeywell™

Catalog No. p-7072290
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6003006 500 g
6003005 1 kg
6003007 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6003006 Supplier Honeywell Chemicals Supplier No. A5132500G
Only null left

Reagent Grade,≥99.0%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name diazanium;sulfate
SMILES N.N.OS(O)(=O)=O

Specifications

Quantity 500 g
Linear Formula (NH4)2SO4
UN Number NONH for all modes of transport
Formula Weight 132.14g/mol
Percent Purity ≥99%
Density 1.77g/mL at 25°C
Chemical Name or Material Ammonium sulfate
RTECSNumber BS4500000