missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfate, 99%, pure

Catalog No. AC446450050
Change view
Click to view available options
Quantity:
5 kg
25 kg
Packaging:
Fibre drum
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446450050 5 kg Plastic Bottle
AC446450250 25 kg Fibre drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446450050 Supplier Thermo Scientific Chemicals Supplier No. 446450050

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
SMILES N.N.OS(O)(=O)=O

Specifications

Melting Point 280.0°C
Quantity 5 kg
Assay Percent Range 99%
Linear Formula (NH4)2SO4
Merck Index 15,552
Solubility Information Solubility in water: 754g/L. Other solubilities: insoluble in alcohol and acetone
IUPAC Name sulfuric acid diamine
Formula Weight 132.14
Percent Purity 99%
Grade Pure
Packaging Plastic Bottle
Chemical Name or Material Ammonium sulfate
Show More Show Less
EINECSNumber 231-984-1

RUO – Research Use Only