missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, 99.0% (ACS Guaranteed Reagent), MilliporeSigma™

Catalog No. MAX13853
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAX13853 2.5 kg
MAX13855 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAX13853 Supplier Technidata Supplier No. AX13853
Only null left

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O

Specifications

Physical Form Crystalline
Quantity 2.5 kg
Linear Formula (NH4)2SO4
Formula Weight 132.14
Grade ACS
Packaging Poly bottle
Density 1.77g/cm3 (20°C); Bulk Density: 850 kg/m3
Chemical Name or Material Ammonium Sulfate, 99.0%, Suprapur™
Recommended Storage Store without restrictions