missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Succinate, MP Biomedicals

Catalog No. ICN205049
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN205049 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN205049 Supplier MP Biomedicals Inc Supplier No. 0520504980
Only null left

Chemical Identifiers

CAS 2226-88-2
Molecular Formula C4H12N2O4
Molecular Weight (g/mol) 152.15
InChI Key NHJPVZLSLOHJDM-UHFFFAOYSA-N
Synonym ammonium succinate, diammonium succinate, unii-dtz9pec9jl, ammonium succinate 1:x, butanedioic acid, diammonium salt, dtz9pec9jl, succinic acid, ammonium salt, succinic acid diammonium salt, caswell no. 046, butanedioic acid, ammonium salt 1:2
PubChem CID 160625
IUPAC Name diazanium;butanedioate
SMILES C(CC(=O)[O-])C(=O)[O-].[NH4+].[NH4+]

Specifications

Quantity 100 g
Formula Weight 152.15
Chemical Name or Material Ammonium Succinate