missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Solution, 400 g (NH4)2HPO4 + 800 mL Water, Ricca Chemical

Catalog No. 6904
Change view
Click to view available options
Package: Natural Poly; Size: 120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
6904 Package: Natural Poly; Size: 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6904 Supplier Ricca Chemical Company Supplier No. RI6904

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

APHA, EPA for Sulfide Testing

Specifications

CAS 7783-28-0, 7732-18-5
CAS Min % 65.0, 32.68
CAS Max % 68.0, 34.01
Molecular Formula H9N2O4P
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
SMILES [NH4+].[NH4+].OP(=O)([O-])[O-]
IUPAC Name diazanium;hydrogen phosphate
Molecular Weight (g/mol) 132.056
PubChem CID 24540
ChEBI CHEBI:63051
Packaging Natural Poly Bottle
Quantity 120 mL
Concentration or Composition Notes 400 g (NH4)2HPO4 + 800mL Water
Chemical Name or Material Ammonium Phosphate Solution
Show More Show Less