missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Monobasic (Crystalline/HPLC), Fisher Chemical

Catalog No. A685500
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A685500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A685500 Supplier Thermo Fisher Scientific Supplier No. FLA685500
Get 1 Extra Chemical When You Buy 2
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name phosphoric acid amine
SMILES N.OP(O)(O)=O

Specifications

Melting Point 190°C
Color White
pH 3.8 to 4.4
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥98.0%
Linear Formula NH4H2PO4
Formula Weight 115.03
Percent Purity ≥98.0%
Grade Also meets ACS specifications
Identification Pass Test
Packaging Glass Bottle
Decomposition Information >190°C
Chemical Name or Material Ammonium Phosphate Monobasic
Show More Show Less