missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Monobasic (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. p-31002
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A684500 500 g Glass Bottle
A6843 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A684500 Supplier Thermo Fisher Scientific Supplier No. FLA684500
Only null left

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name phosphoric acid amine
SMILES N.OP(O)(O)=O

Specifications

Melting Point 190°C
Color White
pH 3.8 to 4.4
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥98.0 %
Linear Formula NH4H2PO4
Formula Weight 115.03
Percent Purity ≥98.0%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Decomposition Information >190°C
Density 1.8g/mL
Chemical Name or Material Ammonium Phosphate Monobasic
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:0
Instability:0