missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Monobasic, ACS Reagent, Honeywell Fluka™

Catalog No. p-7071879
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002115 500 g
6002114 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002115 Supplier Honeywell Chemicals Supplier No. 216003500G
Only null left

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name azanium;dihydrogen phosphate
SMILES N.OP(O)(O)=O

Specifications

Quantity 500 g
Linear Formula NH4H2PO4
UN Number NONH for all modes of transport
Formula Weight 115.03g/mol
Percent Purity ≥98%
Grade ACS Reagent
Chemical Name or Material Ammonium Phosphate Monobasic
EINECSNumber 231-764-5