missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Phosphate, Monobasic, 99.0% min., ACS, MilliporeSigma™
Specifications
Specifications
| Melting Point | 19°C |
| CAS | 7722-76-1 |
| Quantity | 12 kg |
| Assay Percent Range | 99.0% min. |
| Molecular Formula | H6NO4P |
| Linear Formula | (NH4)H2PO4 |
| MDL Number | MFCD00003396 |
| Synonym | ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate |
| InChI Key | LFVGISIMTYGQHF-UHFFFAOYSA-N |
| SMILES | N.OP(O)(O)=O |
| Show More |
Safety and Handling
| Recommended Storage | Store without restrictions |