missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate, Granular, NF, 96-102%, Spectrum™ Chemical

Catalog No. 18600645
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600645 500 g Poly Bottle
18600646 2.5 kg Poly Pail
18600647 12 kg Poly Pail
18600648 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600645 Supplier Spectrum Chemical Mfg Cor Supplier No. A1244500GM
Only null left

A1244, 7783-28-0, (NH4)2HPO4

Spectrum™ Chemical Ammonium Phosphate, Granular, NF is used a buffering agent excipient. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
IUPAC Name phosphoric acid diamine
SMILES N.N.OP(O)(O)=O

Specifications

pH 7.6-8.2
CAS Max % 100%
Quantity 500 g
Percent Purity 96 to 102%
Grade NF
Packaging Poly Bottle