missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Dibasic, puriss. p.a., Honeywell Fluka™

Catalog No. p-7071749
Change view
Click to view available options
Quantity:
250 g
1 kg
Catalog No. Quantity
6001878 250 g
6001877 1 kg
2 options

Catalog No. 6001878

Supplier: Honeywell Chemicals 09840250G

Only null left

Puriss. p.a.,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
PubChem CID 24540
ChEBI CHEBI:63051
IUPAC Name diazanium;hydrogen phosphate
SMILES N.N.OP(O)(O)=O

Specifications

Quantity 250 g
Linear Formula (NH4)2HPO4
UN Number NONH for all modes of transport
Formula Weight 132.06g/mol
Percent Purity ≥99% (T)
Grade puriss. p.a.
Chemical Name or Material Ammonium Phosphate Dibasic
EINECSNumber 231-987-8