missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Dibasic, puriss. p.a. ACS Reagent, Honeywell Fluka™

Catalog No. p-7071965
Change view
Click to view available options
Quantity:
500 g
1 kg
50 kg
Catalog No. Quantity
6002262 500 g
6002261 1 kg
6002263 50 kg
3 options

Catalog No. 6002262

Supplier: Honeywell Chemicals 30402500G

Only null left

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99% (alkalimetric)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
PubChem CID 24540
ChEBI CHEBI:63051
IUPAC Name diazanium;hydrogen phosphate
SMILES N.N.OP(O)(O)=O

Specifications

Quantity 500 g
Linear Formula (NH4)2HPO4
UN Number NONH for all modes of transport
Formula Weight 132.06g/mol
Percent Purity ≥98% (alkalimetric)
Grade ACS/puriss. p.a.
Chemical Name or Material Ammonium Phosphate Dibasic, Reag. Ph. Eur. reag. ISO
EINECSNumber 231-987-8