missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Dibasic, Purified (Diammonium Hydrogen Phosphate), Reagents

Catalog No. 01337485
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337485 100 g
01337484 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337485 Supplier Labchem Inc Supplier No. C2137500100C2
Only null left

Ammonium Phosphate Dibasic, Purified (Diammonium Hydrogen Phosphate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym Diammonium Hydrogen Phosphate
IUPAC Name phosphoric acid diamine
SMILES N.N.OP(O)(O)=O

Specifications

Melting Point 155°C
Color Colorless/White
Physical Form Solid
Quantity 100 g
Grade Pure
Packaging Glass Bottle
Specific Gravity 1.619
Chemical Name or Material Ammonium Phosphate Dibasic, Purified (Diammonium Hydrogen Phosphate)