missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Dibasic (Crystalline/Certified ACS), Fisher Chemical

Catalog No. A686500
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A686500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A686500 Supplier Thermo Fisher Scientific Supplier No. FLA686500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
PubChem CID 24540
ChEBI CHEBI:63051
IUPAC Name phosphoric acid diamine
SMILES N.N.OP(O)(O)=O

Specifications

Melting Point 235°C
Color White
pH 7.9 to 8.3
Physical Form Crystalline Solid
Quantity 500 g
Assay Percent Range ≥98.0%
Linear Formula (NH4)2HPO4
Formula Weight 132.06
Percent Purity ≥98.0%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Density 1.619g/cm³
Chemical Name or Material Ammonium Phosphate Dibasic
Show More Show Less