missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Phosphate, Dibasic, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Supplier: Avantor J.T.Baker 078405
Specifications
| Ammonium Phosphate | |
| 7783-28-0 | |
| 2.5 kg | |
| MFCD00010891 | |
| MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
| phosphoric acid diamine | |
| 24540 | |
| 132.06 | |
| Poly Bottle |
| Dibasic | |
| Crystals | |
| H9N2O4P | |
| diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
| N.N.OP(O)(O)=O | |
| 132.06 | |
| CHEBI:63051 | |
| BAKER ANALYZED™ ACS Reagent | |
| 1L = 1.62kg |
Chemical Identifiers
| 7783-28-0 | |
| 132.06 | |
| MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
| 24540 | |
| phosphoric acid diamine |
| H9N2O4P | |
| MFCD00010891 | |
| diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
| CHEBI:63051 | |
| N.N.OP(O)(O)=O |