missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Dibasic, Crystal, ACS, 98%, Spectrum™ Chemical

Catalog No. 18608758
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608758 2.5 kg Poly Bottle
18608755 25 g Poly Bottle
18608756 125 g Poly Bottle
18608757 500 g Poly Bottle
18608759 12 kg Poly Pail
18608760 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18608758 Supplier Spectrum Chemical Mfg Cor Supplier No. A12402.5KG
Only null left

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
IUPAC Name phosphoric acid diamine
SMILES N.N.OP(O)(O)=O

Specifications

pH 7.7 to 8.1
CAS Max % 1
Quantity 2.5 kg
Percent Purity 98%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.