missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate, Dibasic, ACS Grade, ≥ 98.0%, LabChem™

Catalog No. LC112851
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC112851 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC112851 Supplier LabChem Supplier No. LC112851

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
PubChem CID 24540
ChEBI CHEBI:63051
IUPAC Name phosphoric acid diamine
SMILES N.N.OP(O)(O)=O

Specifications

Melting Point 155°C
Color White
CAS Max % 100
Physical Form Crystals
Quantity 500 g
Linear Formula (NH4)2HPO4
Solubility Information Soluble in water
Formula Weight 132.06
Percent Purity ≥98.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.619g/mL
Decomposition Information Gaseous Ammonia; Phosphorus oxides
Density 1.619g/mL
Chemical Name or Material Ammonium Phosphate, Dibasic
Show More Show Less
Hazard Statement GHS H Statement
Product is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
EINECSNumber 231-987-8
Recommended Storage Room Temperature