missing translation for 'onlineSavingsMsg'
Learn More

Ammonium phosphate, dibasic, 99%, for biochemistry

Catalog No. p-4440131
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447142500 250 g Plastic bottle
AC447140010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447142500 Supplier Thermo Scientific Chemicals Supplier No. 447142500
Only null left

CAS: 7783-28-0 | H9N2O4P | 132.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
PubChem CID 24540
ChEBI CHEBI:63051
SMILES N.N.OP(O)(O)=O

Specifications

Quantity 250 g
Assay Percent Range 99%
Linear Formula (NH4)2HPO4
Merck Index 15,54
Solubility Information (1 M in water) Clear colorless
IUPAC Name diazanium;hydrogen phosphate
Formula Weight 132.06
Percent Purity 99%
Grade Biochemical
Packaging Plastic bottle
Chemical Name or Material Ammonium phosphate, dibasic
Show More Show Less
Hazard Category Warning
EINECSNumber 231-987-8

RUO – Research Use Only