missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Persulfate, Purified (Ammonium Peroxydisulfate), Reagents

Catalog No. 01337489
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337489 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337489 Supplier Labchem Inc Supplier No. C2136500100C2
Only null left

Ammonium Persulfate, Purified (Ammonium Peroxydisulfate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym Ammonium Peroxydisulfate
IUPAC Name diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120°C to 160°C
Color Colorless/White
Physical Form Solid
Quantity 100 g
Grade Pure
Packaging Glass Bottle
Specific Gravity 1.98
Chemical Name or Material Ammonium Persulfate, Purified (Ammonium Peroxydisulfate)