missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Persulfate, Honeywell Fluka™

Catalog No. p-7071993
Change view
Click to view available options
Quantity:
1 kg
5 kg
6 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000355 1 kg
6000356 5 kg
60045905 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6000355 Supplier Honeywell Chemicals Supplier No. 311171KG
Only null left

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Quantity 1 kg
Linear Formula (NH4)2S2O8
UN Number 1444
Solubility Information Soluble in water
IUPAC Name diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Formula Weight 228.2g/mol
Percent Purity ≥98%
Grade ACS/puriss. p.a.
Residue after Ignition ≤0.05% (as SO4)
Density Vapor Density: 7.9 (vs air)
Chemical Name or Material Ammonium Persulfate, Reag. Ph. Eur.
Show More Show Less
Hazard Statement P220-P261-P280-P305 + P351 + P338-P342 + P311
Precautionary Statement H272-H302-H315-H317-H319-H334-H335
EINECSNumber 231-786-5
RTECSNumber SE0350000