missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Persulfate, ACS Reagent (Ammonium Peroxydisulfate), Reagents
Specifications
Specifications
| Melting Point | 120°C to 160°C |
| Color | Colorless/White |
| Physical Form | Solid |
| Quantity | 100 mL |
| Percent Purity | ≥98% |
| Grade | ACS |
| Packaging | Poly Bottle |
| Specific Gravity | 1.98 |
| Chemical Name or Material | Ammonium Persulfate, ACS Reagent (Ammonium Peroxydisulfate) |