missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium persulfate, 99+%, for molecular biology, DNAse, RNAse and Protease free
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Specifications
| Melting Point | 120.0°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 100 g |
| Assay Percent Range | 99% min. (Iodometry) |
| Linear Formula | (NH4)2S2O8 |
| Fieser | 01,952; 02,348; 03,238; 05,15; 06,20; 12,33 |
| Merck Index | 15, 539 |
| Solubility Information | Solubility in water: 582g/L (20°C), may decompose. |
| Formula Weight | 228.19 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause respiratory irritation. Causes skin irritation. May cause an allergic skin reaction. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Take any precaution to avoid mixing with combustibles. IF ON SKIN: Wash with plenty of soap and water. If skin irritation or rash occurs. |
| EINECSNumber | 231-786-5 |
RUO – Research Use Only