missing translation for 'onlineSavingsMsg'
Learn More

Ammonium persulfate, 98+%, ACS reagent

Catalog No. ac401160020
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
100 g
2 kg
25 kg
5 kg
500 g
Packaging:
Fibre drum
Glass bottle
Plastic bottle
Catalog No. Quantity Packaging
AC401160020 2 kg Glass bottle
AC401160100 10 g Glass bottle
AC401161000 100 g Plastic bottle
AC401165000 500 g Plastic bottle
AC401160050 5 kg Plastic bottle
AC401160250 25 kg Fibre drum
6 options

Catalog No. AC401160020

Supplier: Thermo Scientific Chemicals 401160020

Only null left

Oxidation reagent | CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
IUPAC Name diazanium;sulfonatooxy sulfate
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 2 kg
Assay Percent Range 98+%
Linear Formula (NH4)2S2O8
Fieser 01,952; 02,348; 03,238; 05,15; 06,20; 12,33
Merck Index 15, 539
Formula Weight 228.19
Percent Purity ≥98%
Grade ACS Reagent
Packaging Glass bottle
Residue after Ignition 0.05% max.
Specific Gravity 1.98
Vapor Pressure Negligible
Decomposition Information >120°C
Density 1.9800g/mL
Chemical Name or Material Ammonium persulfate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful if swallowed.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement:
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only