missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium Persulfate, Molecular Biology Reagent, MP Biomedicals™
Ammonium Persulfate, Molecular Biology Reagent is a polymerization catalyst used with TEMED for polyacrylamide gel formation.
Manufacturer: Mp Biomedicals 0219398890
Description
- Ammonium persulfate is a polymerization catalyst used with TEMED for polyacrylamide gel formation.
- APS has also been utilized to study protein-protein interactions via photoinitiated crosslinking chemistry.
- Other applications of APS include its use as a reducer and retarder in photography, the manufacture of aniline dyes, electroplating, and the decolorizing and deodorizing of oils.
Specifications
Ammonium persulfate | |
White | |
500 g | |
MFCD00003390 | |
Soluble in water, 83.5 g/100 g water at 25°C | |
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O | |
228.19 | |
228.18 | |
≥98% | |
951 to 1050 kg/m3 |
7727-54-0 | |
120°C (Decomposition) | |
H8N2O8S2 | |
ammonium peroxydisulfate, ammonium persulfate, ammoniumpersulfate, ccris 1430, diammonium peroxodisulphate, diammonium peroxydisulfate, diammonium peroxydisulphate, diammonium persulfate, persulfate d'ammonium french, unii-22qf6l357f | |
ROOXNKNUYICQNP-UHFFFAOYSA-N | |
diazanium;sulfonatooxy sulfate | |
62648 | |
Crystalline Powder | |
Molecular Biology | |
1.98 g/cm3 at 68°F |
Chemical Identifiers
7727-54-0 | |
228.19 | |
ROOXNKNUYICQNP-UHFFFAOYSA-N | |
62648 | |
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O |
H8N2O8S2 | |
MFCD00003390 | |
ammonium peroxydisulfate, ammonium persulfate, ammoniumpersulfate, ccris 1430, diammonium peroxodisulphate, diammonium peroxydisulfate, diammonium peroxydisulphate, diammonium persulfate, persulfate d'ammonium french, unii-22qf6l357f | |
diazanium;sulfonatooxy sulfate |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.